Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529914.2 | XP_011528216.1 | 7, 8 | 813 | ||
XM_011529917.2 | XP_011528219.1 | 7, 8 | 672 | |||
XM_011529918.1 | XP_011528220.1 | 7, 8 | 813 | |||
XM_017028621.1 | XP_016884110.1 | 7, 8 | 694 | |||
XM_017028622.1 | XP_016884111.1 | 7, 8 | 613 | |||
XM_017028624.1 | XP_016884113.1 | 6, 7 | 776 | |||
XM_017028625.1 | XP_016884114.1 | 2, 3 | 164 | |||
GenBank mRNA | AK098703.1 | - | 7, 8 | 638 | ||
BC030013.1 | - | 7, 8 | 423 | 4823215 | ||
BI459310.1 | - | 6, 7 | 381 | |||
CR456341.1 | - | 6, 7 | 400 | |||
HY002382.1 | - | 8, 9 | 463 |