Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321781.1 | NP_001308710.1 | 2 | 198 | ||
NM_001321782.1 | NP_001308711.1 | 2 | 191 | |||
NM_152516.3 | NP_689729.1 | 2 | 295 | |||
XM_011532558.2 | XP_011530860.1 | 2 | 293 | |||
XM_011532559.2 | XP_011530861.1 | 2 | 246 | |||
XM_017003412.1 | XP_016858901.1 | 2 | 103 | |||
GenBank mRNA | AB178811.1 | - | 2 | 279 | ||
AK223607.1 | - | 2 | 284 | |||
AK294203.1 | - | 2 | 263 | |||
BC009266.2 | - | 2 | 248 | |||
BC022046.1 | - | 2 | 267 | 4046433 | ||
D85433.1 | - | 2 | 195 | |||
DT217730.1 | - | 2 | 197 | |||
HY001060.1 | - | 2 | 295 | |||
HY039942.1 | - | 2 | 198 |