Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANSL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307976.1 | NP_001294905.1 | 6 | 1893 | ||
NM_152519.3 | NP_689732.2 | 6 | 1893 | |||
XM_005246328.4 | XP_005246385.1 | 6 | 2175 | |||
XM_005246329.4 | XP_005246386.1 | 6 | 1881 | |||
XM_005246330.4 | XP_005246387.1 | 6 | 1762 | |||
XM_005246332.4 | XP_005246389.1 | 6 | 2174 | |||
XM_011510706.2 | XP_011509008.1 | 6 | 1800 | |||
XM_011510707.2 | XP_011509009.1 | 6 | 1983 | |||
XM_011510710.2 | XP_011509012.1 | 5 | 632 | |||
XM_017003430.1 | XP_016858919.1 | 6 | 1919 | |||
XM_017003431.1 | XP_016858920.1 | 2 | 4001 | |||
GenBank mRNA | AK056911.1 | - | 6 | 1780 | ||
AK074441.1 | - | 5 | 663 | |||
BC032837.1 | - | 6 | 1863 | 4829661 | ||
BC070141.1 | - | 5 | 725 | |||
BC090854.1 | - | 5 | 1231 | |||
BC126155.1 | - | 6 | 1799 | 8991871 | ||
BC143914.1 | - | 6 | 1799 | 9052432 |