Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302769.1 | NP_001289698.1 | 5, 6 | 797 | ||
NM_057177.6 | NP_476518.4 | 5, 6 | 797 | |||
NM_152526.5 | NP_689739.4 | 5, 6 | 797 | |||
NM_205863.3 | NP_995585.2 | 5, 6 | 797 | |||
XM_011510552.2 | XP_011508854.1 | 8, 9 | 1140 | |||
XM_011510553.2 | XP_011508855.1 | 7, 8 | 5863 | |||
XM_017003283.1 | XP_016858772.1 | 4, 5 | 1087 | |||
XM_017003284.1 | XP_016858773.1 | 5, 6 | 4133 | |||
XM_017003285.1 | XP_016858774.1 | 7, 8 | 5870 | |||
XM_017003286.1 | XP_016858775.1 | 4, 5 | 4029 | |||
XM_017003287.1 | XP_016858776.1 | 7, 8 | 5866 | |||
XM_017003288.1 | XP_016858777.1 | 7, 8 | 5868 | |||
XM_017003289.1 | XP_016858778.1 | 2, 3 | 244 | |||
XM_017003290.1 | XP_016858779.1 | 6, 7 | 744 | |||
XM_017003291.1 | XP_016858780.1 | 5, 6 | 616 | |||
XM_017003292.1 | XP_016858781.1 | 5, 6 | 678 | |||
XM_017003293.1 | XP_016858782.1 | 2, 3 | 182 | |||
XM_017003294.1 | XP_016858783.1 | 7, 8 | 5860 | |||
GenBank mRNA | AB073472.1 | - | 7, 8 | 590 | ||
AB092439.1 | - | 7, 8 | 590 | |||
AF428250.1 | - | 5, 6 | 590 | |||
AF428251.1 | - | 5, 6 | 590 | |||
AF466152.1 | - | 5, 6 | 590 | |||
AK056157.1 | - | 7, 8 | 679 |