Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152527.4 | NP_689740.2 | 4 | 1131 | ||
XM_005246353.3 | XP_005246410.1 | 3 | 987 | |||
XM_011510751.2 | XP_011509053.1 | 4 | 902 | |||
XM_011510752.1 | XP_011509054.1 | 4 | 1131 | |||
XM_017003480.1 | XP_016858969.1 | 4 | 825 | |||
GenBank mRNA | AK055356.1 | - | 2 | 388 | ||
AK123207.1 | - | 5 | 888 | |||
AK292873.1 | - | 4 | 863 | |||
BC034382.1 | - | 4 | 1039 | |||
BC065524.1 | - | 4 | 1035 | 6058847 | ||
BX648181.1 | - | 2 | 299 |