Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152455.3 | NP_689668.3 | 5 | 2174 | ||
XM_006720401.3 | XP_006720464.1 | 6 | 2934 | |||
XM_011521266.2 | XP_011519568.1 | 6 | 2614 | |||
GenBank mRNA | AF525399.1 | - | 5 | 2024 | ||
AK093186.1 | - | 4 | 1720 | |||
AK093377.1 | - | 3 | 1209 | |||
AK122788.1 | - | 5 | 2162 | |||
BC017179.2 | - | 3 | 1040 | |||
BC040682.1 | - | 3 | 1040 | |||
BC043612.1 | - | 5 | 2247 | |||
BC109270.1 | - | 5 | 1813 | 40026738 | ||
BC109271.1 | - | 3 | 857 | 40026744 |