Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143687.2 | NP_001137159.1 | 5, 6 | 569 | ||
NM_152494.3 | NP_689707.2 | 6, 7 | 644 | |||
GenBank mRNA | AK057347.1 | - | 6, 7 | 626 | ||
AK301876.1 | - | 5, 6 | 546 | |||
AK301902.1 | - | 6, 7 | 723 | |||
AK301935.1 | - | 5, 6 | 355 | |||
BC064844.1 | - | 5, 6 | 599 | 30378702 |