Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322302.1 | NP_001309231.1 | 7 | 1763 | ||
NM_001322303.1 | NP_001309232.1 | 6 | 1697 | |||
NM_001322304.1 | NP_001309233.1 | 7 | 1841 | |||
NM_001322305.1 | NP_001309234.1 | 5 | 2135 | |||
NM_152495.1 | NP_689708.1 | 6 | 2183 | |||
NR_136287.1 | 5 | 2070 | ||||
NR_136288.1 | 7 | 1693 | ||||
NR_136289.1 | 8 | 1829 | ||||
NR_136290.1 | 7 | 1763 | ||||
NR_136291.1 | 8 | 1899 | ||||
NR_136292.1 | 7 | 1833 | ||||
NR_136293.1 | 10 | 2069 | ||||
NR_136294.1 | 9 | 2003 | ||||
NR_136295.1 | 8 | 1874 | ||||
NR_136296.1 | 8 | 1899 | ||||
NR_136297.1 | 7 | 1763 | ||||
GenBank mRNA | AF070524.1 | - | 5 | 1389 | ||
AK096312.1 | - | 6 | 2183 | |||
BC022780.1 | - | 6 | 1670 | 3894832 |