Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321781.1 | NP_001308710.1 | 2, 3 | 410 | ||
NM_001321782.1 | NP_001308711.1 | 2, 3 | 403 | |||
NM_152516.3 | NP_689729.1 | 2, 3 | 507 | |||
XM_011532559.2 | XP_011530861.1 | 2, 3 | 458 | |||
XM_017003412.1 | XP_016858901.1 | 2, 3 | 315 | |||
GenBank mRNA | AB178811.1 | - | 2, 3 | 491 | ||
AK223607.1 | - | 2, 3 | 496 | |||
BC009266.2 | - | 2, 3 | 460 | |||
BC022046.1 | - | 2, 3 | 479 | 4046433 | ||
D85433.1 | - | 2, 3 | 407 | |||
DT217730.1 | - | 2, 3 | 409 | |||
HY039942.1 | - | 2, 3 | 410 |