Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152536.3 | NP_689749.3 | 18 | 4283 | ||
XM_011533422.1 | XP_011531724.1 | 18 | 4289 | |||
GenBank mRNA | AK096856.1 | - | 7 | 773 | ||
AK097276.1 | - | 18 | 2796 | |||
AK123054.1 | - | 15 | 1475 | |||
AK131078.1 | - | 18 | 3947 | |||
BC035364.1 | - | 15 | 1720 | |||
BC132814.1 | - | 18 | 3921 | 40146836 | ||
BC132816.1 | - | 18 | 3921 | 40146838 | ||
BX538312.1 | - | 18 | 2424 | |||
BX640820.1 | - | 18 | 3224 |