Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTNL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308245.1 | NP_001295174.1 | 3 | 356 | ||
NM_152547.4 | NP_689760.2 | 3 | 356 | |||
XM_006714823.3 | XP_006714886.1 | 3 | 352 | |||
XM_006714824.3 | XP_006714887.1 | 3 | 352 | |||
XM_006714825.3 | XP_006714888.1 | 4 | 743 | |||
XM_011534444.2 | XP_011532746.1 | 3 | 352 | |||
XM_017009113.1 | XP_016864602.1 | 3 | 352 | |||
XM_017009114.1 | XP_016864603.1 | 3 | 223 | |||
XM_017009115.1 | XP_016864604.1 | 3 | 352 | |||
GenBank mRNA | AK057097.1 | - | 3 | 344 | ||
AK298113.1 | - | 2 | 183 | |||
AY358358.1 | - | 3 | 342 | |||
BC062459.1 | - | 3 | 215 | 6185476 | ||
DC379841.1 | - | 3 | 356 |