Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBSCR27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152559.2 | NP_689772.2 | 6 | 773 | ||
XM_017011777.1 | XP_016867266.1 | 6 | 853 | |||
XM_017011778.1 | XP_016867267.1 | 6 | 975 | |||
GenBank mRNA | AF534110.1 | - | 6 | 784 | ||
AY354928.1 | - | 6 | 783 | |||
BC030295.2 | - | 5 | 737 | 5396423 |