Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP54 SIRNA Assays ›
Gene Symbol:
USP54
Gene Name:
ubiquitin specific peptidase 54
Gene Aliases:
C10orf29, bA137L10.3, bA137L10.4
Chromosome Location:
Chr.10: 73497538 - 73626844 on Build GRCh38
UniGene ID:
Hs.657355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
39172
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP54
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320437.1
NP_001307366.1
18
3222
NM_152586.3
NP_689799.3
18
3320
NR_135249.1
19
3707
NR_135250.1
14
2702
XM_005269582.2
XP_005269639.1
17
6398
XM_011539368.2
XP_011537670.1
18
6464
XM_017015759.1
XP_016871248.1
20
5893
XM_017015760.1
XP_016871249.1
20
4795
XM_017015761.1
XP_016871250.1
20
5633
XM_017015762.1
XP_016871251.1
20
4803
XM_017015763.1
XP_016871252.1
20
4097
XM_017015764.1
XP_016871253.1
20
4160
XM_017015765.1
XP_016871254.1
21
4413
XM_017015766.1
XP_016871255.1
20
6019
XM_017015767.1
XP_016871256.1
20
5678
XM_017015768.1
XP_016871257.1
20
3849
XM_017015769.1
XP_016871258.1
20
3457
XM_017015770.1
XP_016871259.1
20
4118
XM_017015771.1
XP_016871260.1
19
6635
XM_017015772.1
XP_016871261.1
19
6641
XM_017015773.1
XP_016871262.1
18
6575
XM_017015774.1
XP_016871263.1
19
5561
XM_017015775.1
XP_016871264.1
20
5439
XM_017015776.1
XP_016871265.1
19
5944
XM_017015777.1
XP_016871266.1
19
5603
XM_017015778.1
XP_016871267.1
18
6521
XM_017015779.1
XP_016871268.1
19
6641
XM_017015780.1
XP_016871269.1
19
6641
XM_017015781.1
XP_016871270.1
18
6470
XM_017015782.1
XP_016871271.1
19
4221
XM_017015783.1
XP_016871272.1
19
5268
XM_017015784.1
XP_016871273.1
18
3214
XM_017015785.1
XP_016871274.1
18
6521
XM_017015786.1
XP_016871275.1
18
3711
GenBank mRNA
AK094637.1
-
1
414
AK126497.1
-
6
1675
AK127213.1
-
5
1255
AK127267.1
-
14
2702
AK303633.1
-
3
913
AL832102.1
-
1
360
BC026037.1
-
1
168
BC064967.1
-
1
356
BC130633.1
-
3
904
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
protein binding
thiol-dependent ubiquitinyl hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0