Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286243.1 | NP_001273172.1 | 2 | 249 | ||
NM_152591.2 | NP_689804.1 | 3 | 354 | |||
XM_006719262.1 | XP_006719325.1 | 4 | 546 | |||
XM_006719263.2 | XP_006719326.1 | 3 | 307 | |||
XM_011537999.1 | XP_011536301.1 | 4 | 597 | |||
XM_011538001.2 | XP_011536303.1 | 4 | 589 | |||
GenBank mRNA | AK093162.1 | - | 3 | 307 | ||
AK302207.1 | - | 2 | 249 | |||
BC044815.1 | - | 3 | 273 | |||
BC073147.1 | - | 2 | 163 | 5742508 | ||
DB036197.1 | - | 3 | 308 |