Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152594.2 | NP_689807.1 | 2 | 493 | ||
XM_005254202.3 | XP_005254259.1 | 3 | 1293 | |||
XM_011521289.2 | XP_011519591.1 | 2 | 190 | |||
GenBank mRNA | AK091222.1 | - | 3 | 263 | ||
AY299089.1 | - | 2 | 493 | |||
BC018015.1 | - | 2 | 489 | |||
BC137480.1 | - | 2 | 1018 | 9021480 | ||
BC137481.1 | - | 2 | 1018 | 9021481 | ||
DA405650.1 | - | 2 | 493 |