Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172225.2 | NP_001165696.1 | 4 | 552 | ||
NM_152606.4 | NP_689819.1 | 4 | 581 | |||
GenBank mRNA | AK122597.1 | - | 4 | 501 | ||
AK290486.1 | - | 4 | 502 | |||
AL832315.1 | - | 4 | 546 | |||
AL832687.1 | - | 4 | 505 | |||
BC017940.1 | - | 4 | 506 | |||
BC126478.1 | - | 4 | 352 | 8992194 | ||
BC126480.1 | - | 4 | 352 | 8992196 | ||
BX537980.1 | - | 4 | 497 |