Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152613.2 | NP_689826.2 | 2 | 217 | ||
GenBank mRNA | AK128138.1 | - | 2 | 217 | ||
AK129656.1 | - | 2 | 170 | |||
BC022546.1 | - | 2 | 154 | 4811804 | ||
BC022549.1 | - | 2 | 154 | 4815849 | ||
CR456400.1 | - | 2 | 130 | |||
DB079211.1 | - | 2 | 217 |