Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145734.2 | NP_001139206.2 | 6 | 845 | ||
NM_001170402.1 | NP_001163873.1 | 6 | 845 | |||
NM_152623.2 | NP_689836.2 | 6 | 845 | |||
GenBank mRNA | AB077043.1 | - | 6 | 814 | ||
AB077044.1 | - | 6 | 683 | |||
AB081750.1 | - | 6 | 814 | |||
AB086378.1 | - | 6 | 814 | |||
AK095246.1 | - | 6 | 843 | |||
BC136572.1 | - | 6 | 715 | 9020562 |