Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152628.3 | NP_689841.3 | 3 | 305 | ||
XM_011516899.1 | XP_011515201.1 | 2 | 282 | |||
XM_017013153.1 | XP_016868642.1 | 3 | 305 | |||
XM_017013154.1 | XP_016868643.1 | 3 | 305 | |||
XM_017013155.1 | XP_016868644.1 | 3 | 305 | |||
XM_017013156.1 | XP_016868645.1 | 3 | 305 | |||
XM_017013157.1 | XP_016868646.1 | 3 | 305 | |||
XM_017013158.1 | XP_016868647.1 | 3 | 305 | |||
XM_017013159.1 | XP_016868648.1 | 3 | 305 | |||
XM_017013160.1 | XP_016868649.1 | 3 | 305 | |||
XM_017013161.1 | XP_016868650.1 | 3 | 305 | |||
XM_017013162.1 | XP_016868651.1 | 3 | 305 | |||
XM_017013164.1 | XP_016868653.1 | 3 | 305 | |||
XM_017013165.1 | XP_016868654.1 | 3 | 305 | |||
GenBank mRNA | AK093906.1 | - | 3 | 305 |