Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018113.2 | NP_001018123.1 | 3 | 440 | ||
NM_001324162.1 | NP_001311091.1 | 3 | 419 | |||
NM_152633.3 | NP_689846.1 | 2 | 319 | |||
NR_136707.1 | 3 | 440 | ||||
XM_011545470.2 | XP_011543772.1 | 3 | 626 | |||
XM_017029355.1 | XP_016884844.1 | 3 | 518 | |||
XM_017029356.1 | XP_016884845.1 | 3 | 626 | |||
GenBank mRNA | AK091383.1 | - | 2 | 319 | ||
BC043596.1 | - | 3 | 423 | |||
BC055411.1 | - | 3 | 433 | |||
BC136558.1 | - | 3 | 422 | 9020548 | ||
BC136560.1 | - | 2 | 301 | 9020550 | ||
BP236844.1 | - | 3 | 433 |