Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000551.3 | NP_000542.1 | 3 | 2366 | ||
NM_198156.2 | NP_937799.1 | 2 | 2243 | |||
GenBank mRNA | BC058831.1 | - | 2 | 2095 | 6457060 | |
BG288777.1 | - | 1 | 553 | |||
BM564781.1 | - | 1 | 160 |