Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000576.2 | NP_000567.1 | 5 | 428 | ||
XM_017003988.1 | XP_016859477.1 | 4 | 429 | |||
GenBank mRNA | AK313983.1 | - | 4 | 1148 | ||
BC008678.1 | - | 5 | 428 | 3875593 | ||
BT007213.1 | - | 4 | 341 | |||
CR407679.1 | - | 4 | 341 | |||
K02770.1 | - | 5 | 427 | |||
M15330.1 | - | 5 | 427 | |||
X02532.1 | - | 5 | 418 | |||
X56087.1 | - | 5 | 398 |