Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152551.3 | NP_689764.3 | 2 | 309 | ||
XM_011514312.2 | XP_011512614.1 | 2 | 245 | |||
GenBank mRNA | AK056796.1 | - | 2 | 286 | ||
AK292409.1 | - | 2 | 287 | |||
AL832462.1 | - | 2 | 310 | |||
BC054008.1 | - | 2 | 282 | |||
BC062458.1 | - | 2 | 271 | |||
BC114630.1 | - | 2 | 277 | 40084896 | ||
BC114631.1 | - | 2 | 278 | 40084903 |