Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168339.1 | NP_001161811.1 | 5 | 534 | ||
NM_001168340.1 | NP_001161812.1 | 5 | 534 | |||
NM_001168341.1 | NP_001161813.1 | 3 | 327 | |||
NM_001168342.1 | NP_001161814.1 | 3 | 409 | |||
NM_152574.2 | NP_689787.2 | 5 | 534 | |||
XM_011517732.2 | XP_011516034.1 | 4 | 331 | |||
XM_017014310.1 | XP_016869799.1 | 7 | 897 | |||
XM_017014311.1 | XP_016869800.1 | 6 | 829 | |||
XM_017014312.1 | XP_016869801.1 | 4 | 399 | |||
GenBank mRNA | AK091187.1 | - | 5 | 420 | ||
AK093459.1 | - | 5 | 601 | |||
AK298581.1 | - | 3 | 182 | |||
AK298998.1 | - | 5 | 506 | |||
AK301185.1 | - | 5 | 385 | |||
AK301427.1 | - | 3 | 325 | |||
AK310271.1 | - | 6 | 392 | |||
AK310482.1 | - | 5 | 534 | |||
BC042417.1 | - | 4 | 401 | |||
BC142985.1 | - | 5 | 513 | 8860428 | ||
DT217817.1 | - | 4 | 506 |