Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152592.3 | NP_689805.3 | 13 | 2352 | ||
XM_005267377.3 | XP_005267434.1 | 14 | 2449 | |||
XM_006720063.3 | XP_006720126.1 | 14 | 2549 | |||
XM_011536515.2 | XP_011534817.1 | 14 | 2484 | |||
XM_017021056.1 | XP_016876545.1 | 13 | 2351 | |||
GenBank mRNA | AK098471.1 | - | 8 | 1115 | ||
AK131436.1 | - | 14 | 2217 | |||
BC146604.1 | - | 14 | 2352 | 40123293 | ||
BX648302.1 | - | 11 | 4224 |