Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000774.4 | NP_000765.2 | 3 | 260 | ||
NR_135528.1 | 3 | 260 | ||||
XM_011526551.2 | XP_011524853.1 | 3 | 400 | |||
XM_011526552.2 | XP_011524854.1 | 3 | 403 | |||
XM_011526553.2 | XP_011524855.1 | 2 | 591 | |||
XM_011526554.2 | XP_011524856.1 | 2 | 218 | |||
XM_011526555.2 | XP_011524857.1 | 2 | 591 | |||
XM_017026384.1 | XP_016881873.1 | 2 | 591 | |||
XM_017026385.1 | XP_016881874.1 | 2 | 591 | |||
XM_017026386.1 | XP_016881875.1 | 2 | 591 | |||
XM_017026387.1 | XP_016881876.1 | 2 | 591 | |||
GenBank mRNA | AK296884.1 | - | 2 | 78 | ||
AK298082.1 | - | 3 | 263 | |||
AK316498.1 | - | 3 | 260 | |||
BC109056.1 | - | 3 | 237 | 40033365 | ||
DC413790.1 | - | 3 | 260 | |||
J02906.1 | - | 3 | 243 |