Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145734.2 | NP_001139206.2 | 7, 8 | 1067 | ||
NM_001170402.1 | NP_001163873.1 | 7, 8 | 1067 | |||
NM_152623.2 | NP_689836.2 | 7, 8 | 1067 | |||
GenBank mRNA | AB077043.1 | - | 7, 8 | 1036 | ||
AB077044.1 | - | 7, 8 | 905 | |||
AB081750.1 | - | 7, 8 | 1036 | |||
AB086378.1 | - | 7, 8 | 1036 | |||
AK095246.1 | - | 7, 8 | 1065 | |||
BC136572.1 | - | 7, 8 | 937 | 9020562 |