Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGEF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300735.1 | NP_001287664.1 | 13 | 1524 | ||
NM_001300736.1 | NP_001287665.1 | 12 | 1401 | |||
NM_152545.2 | NP_689758.1 | 13 | 1527 | |||
XM_017007812.1 | XP_016863301.1 | 11 | 1245 | |||
XM_017007813.1 | XP_016863302.1 | 13 | 1615 | |||
GenBank mRNA | AK056257.1 | - | 13 | 1506 | ||
AK291459.1 | - | 13 | 1472 | |||
AK303625.1 | - | 12 | 1376 | |||
BC121003.1 | - | 13 | 1439 | 40116919 | ||
BC121004.1 | - | 12 | 1316 | 40116920 |