Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152546.2 | NP_689759.2 | 6 | 995 | ||
XM_017009111.1 | XP_016864600.1 | 6 | 1039 | |||
GenBank mRNA | AK058015.1 | - | 6 | 995 | ||
AY611630.2 | - | 6 | 923 | |||
BC017102.1 | - | 1 | 578 | |||
BC031222.1 | - | 6 | 966 | 5297296 |