Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168339.1 | NP_001161811.1 | 10 | 992 | ||
NM_001168340.1 | NP_001161812.1 | 10 | 998 | |||
NM_001168341.1 | NP_001161813.1 | 8 | 791 | |||
NM_001168342.1 | NP_001161814.1 | 8 | 873 | |||
NM_152574.2 | NP_689787.2 | 10 | 998 | |||
XM_011517732.2 | XP_011516034.1 | 9 | 795 | |||
XM_017014310.1 | XP_016869799.1 | 12 | 1361 | |||
XM_017014311.1 | XP_016869800.1 | 11 | 1293 | |||
XM_017014312.1 | XP_016869801.1 | 9 | 863 | |||
GenBank mRNA | AK091187.1 | - | 10 | 884 | ||
AK093459.1 | - | 10 | 1065 | |||
AK298581.1 | - | 8 | 646 | |||
AK298998.1 | - | 10 | 970 | |||
AK301185.1 | - | 9 | 784 | |||
AK301427.1 | - | 8 | 789 | |||
AK310271.1 | - | 11 | 856 | |||
AK310482.1 | - | 10 | 998 | |||
BC042417.1 | - | 9 | 865 | |||
BC142985.1 | - | 10 | 971 | 8860428 |