Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177475.1 | NP_001170946.1 | 3, 4 | 406 | ||
NM_152581.3 | NP_689794.1 | 4, 5 | 442 | |||
XM_005274451.1 | XP_005274508.1 | 4, 5 | 442 | |||
GenBank mRNA | AK093075.1 | - | 3, 4 | 406 | ||
AK315781.1 | - | 4, 5 | 383 | |||
BC030641.1 | - | 4, 5 | 409 | 4821861 | ||
DA708010.1 | - | 4, 5 | 442 |