Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000966.5 | NP_000957.1 | 5, 6 | 971 | ||
NM_001042728.2 | NP_001036193.1 | 3, 4 | 759 | |||
NM_001243730.1 | NP_001230659.1 | 4, 5 | 645 | |||
NM_001243731.1 | NP_001230660.1 | 3, 4 | 496 | |||
NM_001243732.1 | NP_001230661.1 | 2, 3 | 610 | |||
GenBank mRNA | AB307689.1 | - | 3, 4 | 479 | ||
AK290588.1 | - | 5, 6 | 967 | |||
AK297023.1 | - | 4, 5 | 563 | |||
AK297110.1 | - | 3, 4 | 472 | |||
AK297277.1 | - | 2, 3 | 610 | |||
AK297801.1 | - | 3, 4 | 605 | |||
AK301057.1 | - | 3, 4 | 754 | |||
AK316259.1 | - | 3, 4 | 483 | |||
BC093727.1 | - | 3, 4 | 503 | 7939572 | ||
BC093729.1 | - | 3, 4 | 503 | 7939574 | ||
HQ692828.1 | - | 3, 4 | 479 | |||
HQ692829.1 | - | 3, 4 | 446 | |||
L12060.1 | - | 4, 5 | 587 | |||
M24857.1 | - | 5, 6 | 893 | |||
M38258.1 | - | 3, 4 | 625 | |||
M57707.1 | - | 4, 5 | 678 |