Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286441.1 | NP_001273370.1 | 12 | 2133 | ||
NM_152596.3 | NP_689809.2 | 10 | 1878 | |||
XM_006720415.3 | XP_006720478.1 | 11 | 2050 | |||
XM_011521298.2 | XP_011519600.1 | 13 | 2363 | |||
XM_011521299.2 | XP_011519601.1 | 13 | 2094 | |||
XM_011521300.2 | XP_011519602.1 | 10 | 1622 | |||
XM_011521301.2 | XP_011519603.1 | 12 | 1919 | |||
XM_011521302.2 | XP_011519604.1 | 7 | 1436 | |||
XM_017021970.1 | XP_016877459.1 | 10 | 1622 | |||
XM_017021971.1 | XP_016877460.1 | 10 | 1620 | |||
GenBank mRNA | AK123414.1 | - | 2 | 1179 | ||
AK292524.1 | - | 10 | 1784 | |||
AK302864.1 | - | 12 | 2067 | |||
BC030628.2 | - | 10 | 1818 | 4827307 |