Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136257.1 | NP_001129729.1 | 6 | 1821 | ||
NM_001136258.1 | NP_001129730.1 | 7 | 2013 | |||
NM_152621.5 | NP_689834.1 | 6 | 2097 | |||
XM_011531698.2 | XP_011530000.1 | 7 | 2151 | |||
XM_011531699.2 | XP_011530001.1 | 8 | 2025 | |||
XM_011531700.2 | XP_011530002.1 | 7 | 1922 | |||
XM_011531701.2 | XP_011530003.1 | 8 | 3036 | |||
XM_011531702.2 | XP_011530004.1 | 7 | 2964 | |||
XM_017007839.1 | XP_016863328.1 | 7 | 2155 | |||
XM_017007840.1 | XP_016863329.1 | 8 | 2277 | |||
XM_017007841.1 | XP_016863330.1 | 6 | 1984 | |||
GenBank mRNA | AL833599.1 | - | 5 | 1067 | ||
BC028705.1 | - | 6 | 1822 | 4823252 | ||
BC041369.2 | - | 6 | 2090 | 5273650 | ||
BX649142.1 | - | 5 | 1478 |