Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297598.1 | NP_001284527.1 | 9 | 837 | ||
NM_001297599.1 | NP_001284528.1 | 9 | 822 | |||
NM_152622.4 | NP_689835.3 | 9 | 822 | |||
XM_011543216.1 | XP_011541518.1 | 9 | 857 | |||
XM_011543217.1 | XP_011541519.1 | 9 | 857 | |||
XM_017009149.1 | XP_016864638.1 | 10 | 858 | |||
GenBank mRNA | AK093273.1 | - | 7 | 749 | ||
AK299285.1 | - | 8 | 797 | |||
BC041348.2 | - | 9 | 819 | 5272287 | ||
BX537798.1 | - | 9 | 807 | |||
BX640907.1 | - | 9 | 825 | |||
BX647359.1 | - | 9 | 816 | |||
BX648294.1 | - | 9 | 817 | |||
CR749246.1 | - | 10 | 857 | |||
CR749805.1 | - | 10 | 857 |