Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287532.1 | NP_001274461.1 | 3 | 2217 | ||
NM_001287533.1 | NP_001274462.1 | 5 | 2435 | |||
NM_001287534.1 | NP_001274463.1 | 2 | 2090 | |||
NM_007139.3 | NP_009070.2 | 3 | 2186 | |||
NM_152626.3 | NP_689839.1 | 4 | 2313 | |||
GenBank mRNA | AK091618.1 | - | 4 | 2231 | ||
AK093566.1 | - | 1 | 1089 | |||
BC036439.1 | - | 3 | 2109 | 4817733 | ||
BC040594.1 | - | 4 | 2255 | 5269950 | ||
BX647875.1 | - | 2 | 2034 | |||
EU668348.1 | - | 4 | 2257 |