Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042413.1 | NP_001035878.1 | 5 | 2460 | ||
NM_152629.3 | NP_689842.3 | 4 | 1476 | |||
XM_005251386.4 | XP_005251443.1 | 4 | 2019 | |||
XM_005251387.4 | XP_005251444.1 | 4 | 2297 | |||
XM_005251388.4 | XP_005251445.1 | 3 | 1811 | |||
XM_011517763.2 | XP_011516065.1 | 5 | 2139 | |||
XM_011517764.2 | XP_011516066.1 | 5 | 1969 | |||
XM_011517765.2 | XP_011516067.1 | 5 | 2506 | |||
XM_011517766.2 | XP_011516068.1 | 4 | 2548 | |||
XM_011517767.2 | XP_011516069.1 | 4 | 1942 | |||
XM_017014361.1 | XP_016869850.1 | 4 | 2247 | |||
GenBank mRNA | AK096318.1 | - | 2 | 103 | ||
BC033899.2 | - | 4 | 1476 | 4828458 | ||
DQ438877.1 | - | 2341 | ||||
DQ438878.1 | - | 2 | 404 | |||
DQ438879.1 | - | 2 | 287 | |||
DQ438880.1 | - | 2 | 287 | |||
DQ438906.1 | - | 3 | 714 | |||
DQ438907.1 | - | 2 | 111 | |||
KU178885.1 | - | 3 | 1282 | |||
KU178886.1 | - | 3 | 1282 |