Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304548.1 | NP_001291477.1 | 15 | 2757 | ||
NM_152632.3 | NP_689845.2 | 15 | 2757 | |||
XM_017029452.1 | XP_016884941.1 | 15 | 2750 | |||
XM_017029453.1 | XP_016884942.1 | 15 | 2750 | |||
GenBank mRNA | AK126295.1 | - | 15 | 2723 | ||
BC027936.1 | - | 15 | 2612 | 5202137 |