Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152658.2 | NP_689871.1 | 4 | 1401 | ||
XM_005258623.2 | XP_005258680.1 | 4 | 1210 | |||
XM_011526604.2 | XP_011524906.1 | 4 | 1410 | |||
GenBank mRNA | AK057453.1 | - | 5 | 1257 | ||
AK093048.1 | - | 4 | 1402 | |||
AK296640.1 | - | 4 | 895 | |||
AK297630.1 | - | 3 | 711 | |||
BC046207.1 | - | 4 | 950 | 6176484 | ||
BC072416.1 | - | 4 | 1041 | 6169558 | ||
JQ410992.1 | - | 4 | 1278 |