Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152695.5 | NP_689908.3 | 2 | 274 | ||
XM_011531312.2 | XP_011529614.1 | 3 | 344 | |||
XM_011531314.2 | XP_011529616.1 | 2 | 947 | |||
XM_017029350.1 | XP_016884839.1 | 1 | 91 | |||
GenBank mRNA | AK074194.1 | - | 2 | 249 | ||
AK314344.1 | - | 2 | 251 | |||
AL833109.1 | - | 2 | 232 | |||
AL833407.1 | - | 2 | 250 | |||
AY280801.1 | - | 2 | 250 | |||
AY927530.1 | - | 2 | 267 | |||
AY927531.1 | - | 3 | 1079 | |||
BC060768.1 | - | 2 | 249 | 30346713 | ||
BC065938.1 | - | 2 | 237 | |||
DA319333.1 | - | 2 | 274 |