Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152704.3 | NP_689917.2 | 1 | 1810 | ||
NM_199138.2 | NP_954589.1 | 2, 3 | 1144 | |||
XM_005266279.3 | XP_005266336.1 | 2 | 1790 | |||
XM_005266280.3 | XP_005266337.1 | 2 | 1700 | |||
XM_017020415.1 | XP_016875904.1 | 2 | 1464 | |||
GenBank mRNA | AK055049.1 | - | 2, 3 | 1144 | ||
AK098343.1 | - | 1 | 1010 | |||
AK098796.1 | - | 1, 2 | 85 | |||
BC032653.1 | - | 1, 2 | 730 | |||
BC041392.1 | - | 1 | 1819 | 5277030 |