Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145195.1 | NP_001138667.1 | 2 | 511 | ||
NM_001282733.1 | NP_001269662.1 | 2 | 511 | |||
NM_152725.3 | NP_689938.2 | 2 | 511 | |||
GenBank mRNA | AK055061.1 | - | 2 | 459 | ||
AK303735.1 | - | 2 | 196 | |||
BC035118.1 | - | 2 | 443 | |||
BC047635.1 | - | 2 | 426 | 4820920 | ||
BC065917.1 | - | 1 | 255 | |||
BC094700.1 | - | 2 | 387 | 30407123 | ||
BC117323.1 | - | 1 | 261 | 40125874 | ||
BC143551.1 | - | 1 | 260 | 9052062 |