Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286635.1 | NP_001273564.1 | 5 | 787 | ||
NM_001286636.1 | NP_001273565.1 | 6 | 851 | |||
NM_001286637.1 | NP_001273566.1 | 5 | 728 | |||
NM_152734.3 | NP_689947.2 | 4 | 687 | |||
XM_011514377.2 | XP_011512679.1 | 7 | 1550 | |||
XM_017010432.1 | XP_016865921.1 | 7 | 1004 | |||
XM_017010433.1 | XP_016865922.1 | 7 | 1005 | |||
XM_017010434.1 | XP_016865923.1 | 6 | 1243 | |||
GenBank mRNA | AJ420498.1 | - | 4 | 694 | ||
AK058086.1 | - | 6 | 851 | |||
AK172780.1 | - | 4 | 596 | |||
AK300351.1 | - | 5 | 665 | |||
AK315984.1 | - | 5 | 637 | |||
AY358637.1 | - | 5 | 661 | |||
BC023605.2 | - | 4 | 545 | |||
BC095423.1 | - | 5 | 700 | 30348810 | ||
BC105946.1 | - | 4 | 594 | 40032607 | ||
BC105947.1 | - | 4 | 595 | 40032604 | ||
BC107489.1 | - | 4 | 595 | 40032608 | ||
BX538108.1 | - | 4 | 580 | |||
BX647220.1 | - | 5 | 659 |