Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152743.3 | NP_689956.2 | 3 | 554 | ||
XM_005249643.4 | XP_005249700.1 | 3 | 516 | |||
XM_011515177.2 | XP_011513479.1 | 3 | 521 | |||
XM_011515178.1 | XP_011513480.1 | 3 | 732 | |||
XM_011515179.2 | XP_011513481.1 | 3 | 515 | |||
XM_011515181.2 | XP_011513483.1 | 3 | 512 | |||
XM_011515182.2 | XP_011513484.1 | 3 | 307 | |||
XM_011515183.1 | XP_011513485.1 | 3 | 549 | |||
XM_011515184.2 | XP_011513486.1 | 3 | 307 | |||
XM_011515186.2 | XP_011513488.1 | 3 | 513 | |||
XM_017011833.1 | XP_016867322.1 | 3 | 320 | |||
XM_017011834.1 | XP_016867323.1 | 3 | 511 | |||
XM_017011835.1 | XP_016867324.1 | 3 | 549 | |||
XM_017011836.1 | XP_016867325.1 | 3 | 510 | |||
GenBank mRNA | AK024482.1 | - | 3 | 302 | ||
AK131083.1 | - | 3 | 490 | |||
BC015632.2 | - | 3 | 308 | 3839985 | ||
BP363179.1 | - | 3 | 553 |