Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195811.1 | NP_001182740.1 | 3 | 351 | ||
NM_001320272.1 | NP_001307201.1 | 4 | 645 | |||
NM_152666.2 | NP_689879.2 | 4 | 638 | |||
XM_006711752.2 | XP_006711815.1 | 3 | 805 | |||
XM_011544115.2 | XP_011542417.1 | 5 | 713 | |||
XM_011544116.2 | XP_011542418.1 | 5 | 541 | |||
XM_011544120.2 | XP_011542422.1 | 3 | 382 | |||
XM_017000567.1 | XP_016856056.1 | 3 | 818 | |||
XM_017000568.1 | XP_016856057.1 | 3 | 339 | |||
XM_017000569.1 | XP_016856058.1 | 3 | 2343 | |||
GenBank mRNA | AK098092.1 | - | 4 | 645 | ||
AK295421.1 | - | 3 | 332 | |||
AK307857.1 | - | 3 | 330 | |||
AY391836.1 | - | 2 | 201 | |||
AY461578.1 | - | 3 | 351 | |||
BC101373.1 | - | 3 | 278 | 40025860 | ||
BC101374.1 | - | 3 | 278 | 40025861 | ||
BC101375.1 | - | 3 | 278 | 40025862 |