Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011523545.2 | XP_011521847.2 | 20 | 3494 | ||
XM_011523546.2 | XP_011521848.1 | 19 | 3826 | |||
XM_011523548.2 | XP_011521850.1 | 20 | 3522 | |||
XM_011523549.2 | XP_011521851.1 | 19 | 3336 | |||
XM_011523551.2 | XP_011521853.1 | 20 | 3535 | |||
XM_011523552.2 | XP_011521854.1 | 18 | 2894 | |||
XM_011523555.2 | XP_011521857.1 | 17 | 3894 | |||
XM_011523557.2 | XP_011521859.1 | 13 | 2258 | |||
XM_017024462.1 | XP_016879951.1 | 20 | 3460 | |||
XM_017024467.1 | XP_016879956.1 | 20 | 3651 | |||
XM_017024469.1 | XP_016879958.1 | 20 | 3314 | |||
XM_017024470.1 | XP_016879959.1 | 19 | 3115 | |||
XM_017024471.1 | XP_016879960.1 | 19 | 3291 | |||
XM_017024472.1 | XP_016879961.1 | 20 | 3512 | |||
XM_017024475.1 | XP_016879964.1 | 20 | 3221 | |||
XM_017024476.1 | XP_016879965.1 | 18 | 3397 | |||
XM_017024477.1 | XP_016879966.1 | 19 | 3565 | |||
XM_017024478.1 | XP_016879967.1 | 17 | 3028 | |||
GenBank mRNA | AK074334.1 | - | 5 | 819 |