Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KHDRBS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152688.3 | NP_689901.2 | 2 | 413 | ||
XM_011535515.2 | XP_011533817.1 | 2 | 376 | |||
XM_011535516.1 | XP_011533818.1 | 2 | 376 | |||
XM_011535517.1 | XP_011533819.1 | 2 | 376 | |||
XM_011535519.1 | XP_011533821.1 | 2 | 376 | |||
XM_011535520.1 | XP_011533822.1 | 2 | 376 | |||
XM_017010334.1 | XP_016865823.1 | 2 | 376 | |||
XM_017010335.1 | XP_016865824.1 | 2 | 376 | |||
XM_017010336.1 | XP_016865825.1 | 2 | 376 | |||
XM_017010337.1 | XP_016865826.1 | 2 | 376 | |||
XM_017010338.1 | XP_016865827.1 | 2 | 376 | |||
XM_017010339.1 | XP_016865828.1 | 2 | 376 | |||
XM_017010340.1 | XP_016865829.1 | 2 | 376 | |||
XM_017010341.1 | XP_016865830.1 | 2 | 376 | |||
XM_017010342.1 | XP_016865831.1 | 2 | 376 | |||
XM_017010343.1 | XP_016865832.1 | 2 | 376 | |||
XM_017010344.1 | XP_016865833.1 | 2 | 376 | |||
XM_017010345.1 | XP_016865834.1 | 2 | 376 | |||
XM_017010347.1 | XP_016865836.1 | 2 | 299 | |||
XM_017010348.1 | XP_016865837.1 | 2 | 376 | |||
GenBank mRNA | AK095983.1 | - | 2 | 361 | ||
AK292043.1 | - | 2 | 360 | |||
AY077838.1 | - | 2 | 358 | |||
BC034043.1 | - | 2 | 413 | 4796658 |