Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD9L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303496.1 | NP_001290425.1 | 5 | 3791 | ||
NM_001303497.1 | NP_001290426.1 | 6 | 3620 | |||
NM_001303498.1 | NP_001290427.1 | 5 | 3356 | |||
NM_001303500.1 | NP_001290429.1 | 4 | 3224 | |||
NM_152703.3 | NP_689916.2 | 5 | 3996 | |||
XM_005250193.3 | XP_005250250.1 | 6 | 3428 | |||
XM_006715890.1 | XP_006715953.1 | 2 | 2797 | |||
XM_011515903.2 | XP_011514205.1 | 6 | 3681 | |||
XM_017011822.1 | XP_016867311.1 | 4 | 3745 | |||
XM_017011823.1 | XP_016867312.1 | 5 | 3501 | |||
GenBank mRNA | AB095926.1 | - | 5 | 3964 | ||
AF474973.1 | - | 6 | 3373 | |||
AK097204.1 | - | 1 | 177 | |||
AK304307.1 | - | 1 | 2306 | |||
AK304779.1 | - | 1 | 1765 | |||
AL832264.1 | - | 1 | 862 | |||
BC029108.1 | - | 1 | 282 | |||
BC038974.1 | - | 1 | 200 | |||
BC127117.1 | - | 5 | 3232 | 40125349 | ||
BC127118.1 | - | 4 | 3608 | 40125356 |