Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152704.3 | NP_689917.2 | 1 | 1962 | ||
NM_199138.2 | NP_954589.1 | 3 | 1296 | |||
XM_005266279.3 | XP_005266336.1 | 2 | 1942 | |||
XM_005266280.3 | XP_005266337.1 | 2 | 1852 | |||
XM_017020415.1 | XP_016875904.1 | 2 | 1616 | |||
GenBank mRNA | AK055049.1 | - | 3 | 1296 | ||
AK098343.1 | - | 1 | 1162 | |||
AK098796.1 | - | 2 | 237 | |||
BC032653.1 | - | 2 | 882 | |||
BC041392.1 | - | 1 | 1970 | 5277030 |