Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206559.1 | NP_001193488.1 | 3 | 897 | ||
NM_152705.2 | NP_689918.1 | 3 | 730 | |||
XM_005266414.2 | XP_005266471.1 | 4 | 1570 | |||
XM_017020622.1 | XP_016876111.1 | 2 | 597 | |||
XM_017020623.1 | XP_016876112.1 | 4 | 1490 | |||
GenBank mRNA | AF131855.1 | - | 1 | 1483 | ||
AK307356.1 | - | 3 | 694 | |||
BC006972.1 | - | 3 | 630 | |||
BC015319.2 | - | 3 | 639 | 3847651 | ||
BC018528.1 | - | 3 | 643 | 3865616 |